Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPS2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP154663
Description
MRPS2 Polyclonal specifically detects MRPS2 in Human samples. It is validated for Western Blot.Specifications
MRPS2 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
28S ribosomal protein S2, mitochondrial, CGI-91, mitochondrial 28S ribosomal protein S2, mitochondrial ribosomal protein S2, MRP-S2, S2mt | |
Rabbit | |
33 kDa | |
100 μL | |
Core ESC Like Genes, Stem Cell Markers | |
51116 | |
Human | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q9Y399 | |
MRPS2 | |
Synthetic peptides corresponding to MRPS2(mitochondrial ribosomal protein S2) The peptide sequence was selected from the N terminal of MRPS2. Peptide sequence MATSSAALPRILGAGARAPSRWLGFLGKATPRPARPSRRTLGSATALMIR. | |
Affinity purified | |
RUO | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction