Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPS2 Monoclonal Antibody (OTI4D6), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA525318
Description
MRPS2 Monoclonal antibody specifically detects MRPS2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
MRPS2 | |
Monoclonal | |
1.2 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9Y399 | |
MRPS2 | |
Full length human recombinant protein of MRPS2 produced in E.coli | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI4D6 | |
Unconjugated | |
MRPS2 | |
1500019M10Rik; 28S ribosomal protein S2, mitochondrial; CGI-91; mitochondrial ribosomal protein S2; Mitochondrial small ribosomal subunit protein uS2m; MRPS2; MRP-S2; S2mt | |
Mouse | |
Affinity Chromatography | |
RUO | |
51116 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction