Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRPS25 Mouse anti-Human, Alexa Fluor™ 594, Clone: 380000000, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP242628AF594

Catalog No. NP242628A94


Add to cart

Description

Description

MRPS25 Monoclonal antibody specifically detects MRPS25 in Human samples. It is validated for Western Blot,ELISA.
Specifications

Specifications

MRPS25
380000000
Western Blot, ELISA
FLJ00023, mitochondrial 28S ribosomal protein S25, mitochondrial ribosomal protein S25, MRP-S2528S ribosomal protein S25, mitochondrial, RPMS25DKFZp313H0817, S25mt
Mouse
IgG2a Kappa
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
MRPS25
Recombinant human MRPS25 (1-173aa) purified from E. coli
100 μL
Primary
64432
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit