Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ MRPS25 Recombinant Protein
Supplier: Abnova™ H00064432P012
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Antibody Production, ELISA, Protein Array, Western Blot | |
44.8 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
2μg | |
-80°C | |
Recombinant |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
Human MRPS25 Full-length ORF Recombinant Protein with GST-tag at N-terminal | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE stained with Coomassie Blue | |
Wheat Germ (in vitro) | |
Human | |
Solution |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction