Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPS30 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA551245
Description
The antibody detects endogenous levels of total MRPS30 protein.
MRPS30 Polyclonal antibody specifically detects MRPS30 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
MRPS30 | |
Polyclonal | |
Unconjugated | |
Mrps30 | |
2610020A16Rik; 28S ribosomal protein S30, mitochondrial; 39S ribosomal protein S30, mitochondrial; AA968347; BM-047; Mitochondrial large ribosomal subunit protein mL65; Mitochondrial large ribosomal subunit protein mS30; mitochondrial ribosomal protein S30; MRP S30; Mrps30; MRP-S30; PAP; PDCD9; programmed cell death 9; Programmed cell death protein 9; S30mt | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10884 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9NP92 | |
Mrps30 | |
Fusion protein of human MRPS30 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction