Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRPS30 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP287837

Catalog No. NB035459


Add to cart

Description

Description

MRPS30 Polyclonal specifically detects MRPS30 in Human samples. It is validated for Western Blot.
Specifications

Specifications

MRPS30
Unconjugated
PBS, 2% Sucrose
28S ribosomal protein S30, mitochondrial, DKFZp566B2024, mitochondrial ribosomal protein S30, MRP-S30, PAP, PDCD9, programmed cell death 9, Programmed cell death protein 9, S30mt
The immunogen is a synthetic peptide directed towards the C-terminal region of human MRPS30
100 μg
Apoptosis, Core ESC Like Genes, Stem Cell Markers
Primary
10884
Western Blot
Western Blot 1.0 ug/ml
Purified
Rabbit
Immunogen affinity purified
RUO
Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit