Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPS34 Mouse anti-Human, Mouse, Rat, Biotin, Clone: OTI4H2, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272788B
Description
MRPS34 Monoclonal specifically detects MRPS34 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
MRPS34 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
MRPS34 | |
Full length recombinant protein of human MRPS34 (NP_076425) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI4H2 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
MGC2616, mitochondrial ribosomal protein S34, MRPS12, MRP-S12,28S ribosomal protein S34, mitochondrial, MRP-S34, S34mt | |
Mouse | |
Affinity Purified | |
Stem Cell Markers | |
65993.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title