Learn More
MRRF Mouse anti-Human, Clone: OTI1B8, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF806216
Catalog No. 50-166-7739
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
MRRF is responsible for the release of ribosomes from messenger RNA at the termination of protein biosynthesis, may increase the efficiency of translation by recycling ribosomes from one round of translation to another.
Specifications
Specifications
MRRF
Monoclonal
Unconjugated
MRRF
Lyophilized
MRRF
Human recombit protein fragment corresponding to amino acids 56-262 of human MRRF produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI1B8
PBS with 8% trehalose and no preservative; pH 7.3
Q96E11
MRFF, MTRRF, RRF
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
92399