Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris™ MRS 2578

Selective P2Y6 antagonist

Manufacturer:  Tocris™ 2146/10

 View more versions of this product

Catalog No. 21-461-0R


  Check Availability
Add to cart

Disclaimers:

For Research Use Only


Specifications

Description

Selective antagonist of P2Y6 nucleotide receptors; IC50 values are 37 and 98nM at human and rat P2Y6 receptors respectively. Displays no activity at P2Y1, P2Y2, P2Y4 and P2Y11 receptors (IC50 > 10µM). Inhibits agonist-induced cardiomyocyte contraction and UDP-induced phagocytosis.
  • Chemical Name: N,N''-1,4-Butanediylbis[N'-(3-isothiocyanatophenyl)thiourea
  • Formula: C20H20N6S4
  • CAS Number: 711019-86-2
  • Solubility: Soluble to 50mM in DMSO
Description & Specifications

Specifications

MRS 2578
MRS 2578
472.66
10mg
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit