Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRT 10, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
330829-30-6
C24H23N3O5S
465.524
KVQVEZQDNHMQJV-UHFFFAOYSA-N
mrt 10,mrt-10,aronis27184,mrt hplc,n-3-3-3,4,5-trimethoxy-benzoyl-thioureido-phenyl-benzamide,n-3-benzoylamino phenyl amino carbonothioyl-3,4,5-trimethoxybenzamide,n-3-benzoylamino phenyl amino thioxomethyl-3,4,5-trimethoxybenzamide,n-3-3,4,5-trimethoxybenzoyl amino carbothioyl amino phenyl benzamide,3,4,5-trimethoxy-n-3-phenylcarbonyl amino phenyl carbamothioyl benzamide,n-3-benzoyl amino phenyl amino thioxo methyl-3,4,5-trimethoxybenzamide
1139102
N-[(3-benzamidophenyl)carbamothioyl]-3,4,5-trimethoxybenzamide
COC1=CC(=CC(=C1OC)OC)C(=O)NC(=S)NC2=CC=CC(=C2)NC(=O)C3=CC=CC=C3
This item is not returnable. View return policy
330829-30-6
C24H23N3O5S
465.524
KVQVEZQDNHMQJV-UHFFFAOYSA-N
mrt 10,mrt-10,aronis27184,mrt hplc,n-3-3-3,4,5-trimethoxy-benzoyl-thioureido-phenyl-benzamide,n-3-benzoylamino phenyl amino carbonothioyl-3,4,5-trimethoxybenzamide,n-3-benzoylamino phenyl amino thioxomethyl-3,4,5-trimethoxybenzamide,n-3-3,4,5-trimethoxybenzoyl amino carbothioyl amino phenyl benzamide,3,4,5-trimethoxy-n-3-phenylcarbonyl amino phenyl carbamothioyl benzamide,n-3-benzoyl amino phenyl amino thioxo methyl-3,4,5-trimethoxybenzamide
1139102
N-[(3-benzamidophenyl)carbamothioyl]-3,4,5-trimethoxybenzamide
COC1=CC(=CC(=C1OC)OC)C(=O)NC(=S)NC2=CC=CC(=C2)NC(=O)C3=CC=CC=C3

Smoothened (Smo) antagonist

Smoothened (Smo) receptor antagonist (IC50 = 0.5 μM in HEK293 cells transiently expressing mouse Smo). Inhibits ShhN-induced Gli luciferase reporter activity in Shh-light 2 cells (IC50 = 2.5 μM); acts as an inverse agonist during nontranscriptional Hedgehog pathway activation (IC50 = 2.5 μM in HEK293 cells).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 465.52
Percent Purity >98%
Chemical Name or Material MRT 10

Recommended Storage : Store at +4°C