Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MRTO4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580827
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human MRTO4/MRT4.
MRTO4 is a protein sharing a low level of sequence similarity with ribosomal protein P0. While the precise function of the protein is currently unknown, it appears to be involved in mRNA turnover and ribosome assembly.
Specifications
MRTO4 | |
Polyclonal | |
Unconjugated | |
MRTO4 | |
2610012O22Rik; 60S acidic ribosomal protein PO; C1orf33; dJ657E11.4; fb71a02; hypothetical protein LOC550350; Mg684; mRNA turnover 4 homolog; mRNA turnover 4 homolog (S. cerevisiae); mRNA turnover protein 4 homolog; MRT4; MRT4 homolog, ribosome maturation factor; MRT4, mRNA turnover 4, homolog; MRT4, mRNA turnover 4, homolog (S. cerevisiae); mrto4; MRTO4 ribosome maturation factor; muscle protein684; RGD1311709; Ribosome assembly factor MRTO4; wu:fb71a02; zgc:110388 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
51154 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with no preservative | |
Q9UKD2 | |
MRTO4 | |
Recombinant Human MRTO4/MRT4 protein (Met1-Asp239). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction