Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD11b Rat anti-Mouse, BUV395, Clone: M1/70, BD Horizon™
Rat Monoclonal Antibody
Supplier: BD Biosciences 565976
Description
CD11b Monoclonal antibody specifically detects CD11b in Mouse samples. It is validated for Flow cytometry.Specifications
CD11b | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Rat | |
25 μg | |
Cell Biology | |
16409 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity and unconjugated antibody and free BD Horizon™ BUV395 were removed. chromatography. The antibody was conjugated with BD Horizon™ BUV395 under optimum conditions | |
IgG2b κ |
Flow Cytometry | |
M1/70 | |
Brilliant Ultraviolet 395 | |
Itgam; Integrin alpha-M; Ly-40; Mac-1a; Mac-1 alpha; CR3A; CR-3 alpha chain | |
Mouse Splenic Cells | |
RUO | |
Primary | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?
Provide Content Correction