Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD272 Hamster anti-Mouse, Brilliant Violet 421, Clone: HMBT-6B2, BD Optibuild™

Hamster Monoclonal Antibody

Manufacturer:  BD Biosciences 740099

Catalog No. BDB740099


Add to cart

Description

Description

The HMBT-6B2 monoclonal antibody specifically binds to CD272 which is also known as B- and T-lymphocyte attenuator (BTLA). CD272 is a type 1 transmembrane glycoprotein and a member of the Ig superfamily. CD272 is variably expressed on developing and mature T and B lymphocytes, NKT cells, NK cells, macrophages and dendritic cells. CD272 expression is upregulated by activated T cells including Th1, Th2, and T follicular helper cells and by anergic T cells. CD272 is structurally similar to CD152/CTLA-4 and CD279/PD-1 and functions as a coinhibitory receptor for B and T cell responses. Herpesvirus entry mediator (HVEM), also known as CD270 and LIGHT-R, has been identified as a ligand for CD272. The crosslinking of CD272 by HVEM inhibits T-cell proliferation and cytokine production.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

Specifications

Specifications

CD272
HMBT-6B2
Brilliant Violet 421
Aqueous buffered solution containing ≤0.09% sodium azide.
Hamster
IgG
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Btla; B and T lymphocyte attenuator; B- and T-lymphocyte-associated protein
Affinity Purified
Mouse CD272
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only