Learn More
CD45RC Rat anti-Mouse, BB700, Clone: DNL-1.9, BD Optibuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 742087
Description
The DNL-1.9 antibody monoclonal antibody specifically recognizes an exon C-dependent epitope of CD45 protein which is found at high density on B cells, and at low density on peripheral T cytotoxic/suppressor cells, a subset of peripheral T helper cells, and a very small subset of thymocytes. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: Its intracellular (COOHterminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C, respectively), plus differing levels of glycosylation. The CD45 isoforms detected in the mouse are cell type-, maturation-, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction
The antibody was conjugated to BD Horizon™ BB700, which is part of the BD Horizon Brilliant™ Blue family of dyes. It is a polymer-based tandem dye developed exclusively by BD Biosciences. With an excitation max of 485 nm and an emission max of 693 nm, BD Horizon BB700 can be excited by the 488 nm laser and detected in a standard PerCP-Cy™5.5 set (eg, 695/40nm filter). This dye provides a much brighter alternative to PerCP-Cy5.5 with less cross laser excitation off the 405 nm and 355 nm lasers.BD Horizon Brilliant™ Blue 700 (BB700) is a dye that was exclusively developed by BD Biosciences as brighter alternative to PerCP-Cy5.5. This dye also has less cross laser excitation off the 405 nm laser, resulting in less spillover into the violet channels compared to PerCP-Cy5.5. Due to similar excitation and emission properties, BD Horizon BB700 and PerCP-Cy5.5 cannot be used simultaneously.Specifications
CD45RC | |
DNL-1.9 | |
Brilliant Blue 700 (BB700) | |
Ptprc, Cd45, CD45R, L-CA, Ly-5, Lyt-4, T200 | |
70Z/3 mouse pre-B cell line | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.