Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49d Rat anti-Mouse, Brilliant Violet 421, Clone: 9C10(MFR4.B), BD Optibuild™

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 740016

Catalog No. BDB740016


Add to cart

Description

Description

The 9C10 (MFR4.B) monoclonal antibody specifically binds to the Integrin α 4 chain (CD49d), which is expressed as a heterodimer with either of two β chains, β 1 or β 7. The α 4β 1 integrin (VLA-4, CD49d/CD29) is expressed on most peripheral lymphocytes, thymocytes, and monocytes; while the α β 7 also preferentially binds to the mucosal vascular addressin, MAdCAM-1. Although the 9C10 (MFR4.B) antibody alone has been reported to have little function-blocking activity, it can augment the inhibitory effects of mAb R1-2 (Cat. No. 553153), resulting in almost complete inhibition of VLA-4 binding to VCAM-1.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

Specifications

Specifications

CD49d
9C10(MFR4.B)
Brilliant Violet 421
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2a, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Integrin α4; Itga4; Integrin alpha-4; LPAM alpha; VLA-4 alpha; VLA-4a
Affinity Purified
Mouse (C57BL/6xA/J)F[1] Fetal Liver Mast MC/9
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only