Learn More
CD72 a, b, and d Alloantigens Mouse anti-Mouse, Brilliant Violet 786, Clone: K10.6, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 740940
Description
The K10.6 monoclonal antibody specifically recognizes Lyb-2.1, Lyb-2.2, and Lyb-2.4 (CD72 a, b, and d alloantigens, respectively). CD72 is a 45-kDa type-II membrane protein, containing a C-type lectin-like domain and ITIM and ITIM-like sequences in the cytoplasmic tail. CD72 is expressed at all stages of B-lymphocyte development except plasma cells, and it has been shown to negatively regulate B-cell receptor signaling. Analysis of CD72-deficient mice supports these results and shows that CD72 is involved in B-cell development. CD72-stimulated B cells show a transient association of CD19 with CD72, as well as an increase in Tyr-phosphorylation of CD19. CD72 is reported to be a ligand for CD5, although this is controversial. It is also reported to be a ligand for CD100 (Sema4D). The CD72 alloantigens Lyb-2.1 (originally identified as Ly-m19.2), Lyb-2.2 (originally identified as Ly-32.2), Lyb-2.3, and Lyb-2.4 are encoded by the Cd72[a], Cd72[b], Cd72[c], and Cd72[d] alleles, respectively. Lyb-2.1 is expressed on B lymphocytes of CBA/J, C3H/Bi, C57BR, C57L, C58, DBA/1, DBA/2, and SWR strains. Lyb-2.2 is expressed on B lymphocytes, a subset of peripheral T cells, and activated T lymphocytes in A, BALB/c, CBA/H, C3H/He, C57BL, PL, and 129 strains. Lyb-2.4 is expressed on a subset of splenocytes of the STS/A strain. K10.6 antibody does not react with Lyb-2.3 (AKR and SJL strains) nor with non-lymphoid tissues. Five serological specificities of CD72 alloantigens have been described and the nomenclature CD72.1, CD72.2, CD72.3, CD72.4, and CD72.5 proposed, which does not correspond to the names of the alleles. Some authors have referred to the specificity of mAb K10.6 as CD72.4.
The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60-nm filter).
Specifications
CD72 a, b, and d Alloantigens | |
Monoclonal | |
0.2mg/mL | |
Lyb-2.1, Lyb-2.2, and Lyb-2.4 | |
Mouse | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2b κ |
Flow Cytometry | |
K10.6 | |
Brilliant Violet 786 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
DBA/2)F1 hybrid mouse pre-B-cell leukemia line 70Z/3, (C57BL/6 | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only