Learn More
CD93 (Early B Lineage) Rat anti-Mouse, Brilliant Violet 605, Clone: AA4.1, BD Optibuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 740386
Description
The AA4.1 monoclonal antibody specifically recognizes the Early B Lineage antigen which is also known as CD93, AA4 antigen, Ly-68, and Complement component C1q receptor (C1qRp). This 130-140-kDa type I transmembrane glycoprotein is expressed on immature B lymphocytes in the adult bone marrow and on hematopoietic progenitors and stem cells in adult bone marrow, fetal liver, and embryonic yolk sac. Although CD93+ cells are most plentiful in adult mouse bone marrow, a smaller number of CD93+ cells which express lower CD93 levels can be detected in the adult spleen using bright fluorescent conjugates of the AA4.1 antibody or an amplified indirect immunofluorescent staining procedure. It has been observed that the staining pattern of the 493 monoclonal antibody is similar to that of the AA4.1 antibody, that both antibodies precipitate molecules of the same molecular weight, and that staining with the AA4.1 antibody is not blocked by the 493 antibody. These results suggest that the antibodies recognize separate epitopes on the same Early B Lineage antigen.
This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max of 602-nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20-nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605-nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser.
Specifications
CD93 (Early B Lineage) | |
Monoclonal | |
0.2mg/mL | |
Aa4; Cd93; C1qRp; C1qr1; Ly-68; Ly68; C1q/MBL/SPA receptor | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2b κ |
Flow Cytometry | |
AA4.1 | |
Brilliant Violet 605 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Pre-B lymphoma 70Z/3, derived from (C57BL/6 x DBA/2)F1 mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only