Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD93 (Early B Lineage) Rat anti-Mouse, BUV496, Clone: AA4.1, BD Biosciences™
Rat Monoclonal Antibody
Supplier: BD Biosciences 741124
Description
CD93 (Early B Lineage) Monoclonal antibody specifically detects CD93 (Early B Lineage) in Mouse samples. It is validated for Flow CytometrySpecifications
CD93 (Early B Lineage) | |
Monoclonal | |
Brilliant Ultraviolet 496 | |
Aa4; Cd93; C1qRp; C1qr1; Ly-68; Ly68; C1q/MBL/SPA receptor | |
Pre-B lymphoma 70Z/3, derived from (C57BL/6 x DBA/2)F1 mouse | |
RUO | |
Mouse | |
IgG2b κ |
Flow Cytometry | |
AA4.1 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV496 under optimal conditions that minimize unconjugated dye and antibody. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction