Learn More
Laxco™ MS Series Fixed Magnification Stereo Microscope System
Utilizes fixed dual magnification optical system. Laxco™ MS Series Fixed Magnification Stereo Microscope System are research-grade instrument with wide choice of zoom magnification ranges and most commonly found in general laboratory.
Manufacturer:  Laxco™ MS10215
 View more versions of this product
Catalog No. MS10215
Add to cart
Description
Description
Optics
  • Choose between multiple combinations of magnifications
  • Dual diopter adjustments accommodates different eye focal length
  • Parfocality maintained between magnification
  • Clearly marked neural knobs for magnification
Stand
  • Rugged, all metal construction
  • Large, contoured stage
  • Left and right hand operation
  • Slip clutch prevents damage to the focusing mechanism
  • Locked-in stage clip
Light Source
  • 20w halogen lower illumination
  • 15w halogen, with reflector for upper illumination
  • Individually control light sources
Eyepiece
  • 10X super-widefield eyepieces with interpupillary and diopter adjustments to correct for user-specific eyesight
Certifications
Quality Proformance Certification
Specifications
Specifications
15X to 30X
Stereo Microscope
15X
2 Year - Electronic, Mechanical, Optical with option to extend to 5 years
Transmitted Fluorescent and Halogen Reflected