Learn More
TCR β Chain Hamster anti-Mouse, BUV395, Clone: H57-597, BD Optibuild™
Armenian Hamster Monoclonal Antibody
Supplier: BD Biosciences 742485
Description
The H57-597 antibody reacts with a common epitope of theβchain of the T-cell Receptor (TCR) complex onαβTCR-expressing thymocytes, peripheral T lymphocytes, NK1.1+ thymocytes, and NK-T cells of all mouse strains tested. It does not react withγδTCR-bearing T cells. In the fetal and adult thymus, the TCRβ-chain may form homodimers or pair with the pre-TCRα-chain on the surface of immature thymocytes before TCRα-chain expression. Plate-bound or soluble H57-597 antibody activatesαβTCR-bearing T cells, and plate-bound mAb can induce apoptotic death.
The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.Specifications
TCR β Chain | |
H57-597 | |
Brilliant Ultraviolet 395 | |
Tcrb, TCRbeta, TCRβ, T cell receptor beta chain | |
TCR affinity-purified from mouse T-cell hybridoma DO-11.10 | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Armenian Hamster | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2 λ1 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.