Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vγ1.1 TCR Armenian Hamster anti-Mouse, BV650, Clone: 2.11, BD OptiBuild
Armenian Hamster Monoclonal Antibody
Manufacturer: BD Biosciences 745310
Description
Vγ1.1 TCR Monoclonal antibody specifically detects Vγ1.1 TCR in Mouse samples. It is validated for Flow Cytometry.Specifications
Vγ1.1 TCR | |
2.11 | |
Brilliant Violet 650 | |
Armenian Hamster | |
50 μg | |
Refrigerate (2 to 8°C) | |
Monoclonal |
Flow Cytometry | |
0.2 mg/mL | |
Purified | |
IgG | |
RUO | |
Primary | |
Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title