Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MS049 oxalate salt

Potent and selective PRMT4 and PRMT6 inhibitor

$147.00 - $590.00

Specifications

Chemical Name or Material N-Methyl-4-(phenylmethoxy)-1-piperidineethanamine oxalate salt
Molecular Formula C15H24N2O.C2H2O4
Product Type Target: Histone Methyltransferase Inhibitors
Formula Weight 338.4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
56-851-0 View Documents Tocris Bioscience™
5685/10
10mg Each for $147.00
Add to cart
 
56-855-0 View Documents Tocris Bioscience™
5685/50
50mg Each for $590.00
Add to cart
 
Description

Description

Potent and selective PRMT4 and PRMT6 inhibitor (IC50 values are 44 and 63 nM, respectively). Reduces Med12me2a and H3R2me2a levels in HEK293 cells in vitro.

To request the negative control for MS049, please fill out the MS049N request form on the SGC website.

Specifications

Specifications

N-Methyl-4-(phenylmethoxy)-1-piperidineethanamine oxalate salt
C15H24N2O.C2H2O4
Target: Histone Methyltransferase Inhibitors
338.4
Safety and Handling

Safety and Handling

Recommended Storage : Desiccate at RT

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit