Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MS4A1/CD20 Mouse anti-Human, Alexa Fluor 350, Clone: 396444, R&D Systems™

Mouse Monoclonal Antibody

Manufacturer:  R&D Systems FAB4225U100UG

Catalog No. FAB4225U100


Add to cart
This item is not returnable. View return policy

Description

Description

CD20 Monoclonal specifically detects CD20 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD20
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
P11836
MS4A1
NS0 mouse myeloma cell line transfected with human MS4A1/CD20 Met1-Pro297 Accession # P11836
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human MS4A1/CD20. Stains human MS4A1/CD20 transfectants but not irrelevant transfectants.
Flow Cytometry
396444
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35, Bp35MGC3969, CD20, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Ly-44, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A1, MS4A2, S7
Mouse
IgG1
RUO
Primary
931
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)