Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MS4A10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553233
Description
Immunogen sequence: KAEATVIPSR CARGLPSWQV LSPVQPWQTS APQNTTQPKL LAPHQHEKSQ KKSSLLKE Highest antigen sequence identity to the following orthologs: Mouse - 32%, Rat - 30%.

Specifications
MS4A10 | |
Polyclonal | |
Unconjugated | |
MS4A10 | |
CD20 antigen-like 7; CD20L7; membrane spanning 4-domains A10; membrane-spanning 4-domains subfamily A member 10; membrane-spanning 4-domains, subfamily A, member 10; MS4A10; MS4A9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
341116 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.08 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96PG2 | |
MS4A10 | |
Recombinant protein corresponding to Human MS4A10. Recombinant protein control fragment (Product #RP-91110). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction