Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MS4A14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5139841
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: DQLQFVLQEE FSSDDSTTNA QSVIFGGYAF FKLTLSRSPL VSQPGNKGRE FVPDEQKQSI LPSPKFSEEE IEPLPP.
May be involved in signal transduction as a component of a multimeric receptor complex.
Specifications
MS4A14 | |
Polyclonal | |
Unconjugated | |
MS4A14 | |
membrane spanning 4-domains A14; membrane-spanning 4-domains subfamily A member 14; membrane-spanning 4-domains, subfamily A, member 14; membrane-spanning 4-domains, subfamily A, member 16; MS4A13; MS4A13 protein; MS4A14; MS4A16; NYD-SP21; testes development-related NYD-SP21; testis development protein NYD-SP21 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84689 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96JA4 | |
MS4A14 | |
Recombinant protein corresponding to Human MS4A14. Recombinant protein control fragment (Product #RP-108964). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction