Learn More
MS4A3 Mouse anti-Human, Alexa Fluor 647, Clone: 489433, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB6247R100UG
Catalog No. FAB6247R100
Add to cart
This item is not returnable. View return policy
Description
Description
MS4A3 Monoclonal specifically detects MS4A3 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
MS4A3
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
NP_006129
MS4A3
NS0 mouse myeloma cell line transfected with human MS4A3 Accession # NP_006129
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human MS4A3 in direct ELISAs.
Flow Cytometry
489433
Alexa Fluor 647
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
CD20 antigen homolog, CD20 antigen-like protein, CD20L, CD20LIgE receptor beta subunit, hematopoietic cell 4 transmembrane protein, Hematopoietic-specific transmembrane protein 4, HTM4, HTM4IgE receptor beta chain, membrane-spanning 4-domains subfamily A member 3, membrane-spanning 4-domains, subfamily A, member 3 (hematopoieticcell-specific)
Mouse
IgG2a
RUO
Primary
932
Human
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)