Learn More
MS4A4A 293T Cell Overexpression Lysate 2 (Denatured), Abnova
Manufacturer:  Abnova Corporation H00051338T02
Catalog No. 89-002-222
Add to cart
Description
Description
Western Blotting
Specifications
Specifications
Western Blot
293T
NM_024021.2
6.8
Transient overexpression cell Lysate was tested with Anti-MS4A4A antibody (H00051338-B02) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
MS4A4A 293T Cell Transient Overexpression Lysate(Denatured)
23.2
pCMV-MS4A4A full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.