Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MS4A8B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548952

Catalog No. PA548952

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May be involved in signal transduction as a component of a multimeric receptor complex.
Specifications

Specifications

MS4A8B
Polyclonal
PBS with 0.09% sodium azide
Q9BY19
Membrane-spanning 4-domains subfamily A member 8, Four-span transmembrane protein 4, Membrane-spanning 4-domains subfamily A member 8B, MS4A8, 4SPAN4, MS4A8B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
83661
Western Blot
Unconjugated
MS4A8
Liquid
MS4A8
KLH conjugated synthetic peptide between 44-70 amino acids from the N-terminal region of human MS4A8B.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit