Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MS4A8B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552420
Description
Immunogen sequence: EFGIACASSH FGCQLVCCQS SNVSVIYPNI YAANPVITPE PVTSPPSYSS EIQANK Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 52%.
May be involved in signal transduction as a component of a multimeric receptor complex.
Specifications
MS4A8B | |
Polyclonal | |
Unconjugated | |
MS4A8 | |
4SPAN4; CD20L5; Four-span transmembrane protein 4; membrane spanning 4-domains A8; membrane-spanning 4-domains subfamily A member 8; Membrane-spanning 4-domains subfamily A member 8B; membrane-spanning 4-domains, subfamily A, member 8; membrane-spanning 4-domains, subfamily A, member 8A; membrane-spanning 4-domains, subfamily A, member 8B; MS4A4; MS4A8; Ms4a8a; MS4A8B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83661 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BY19 | |
MS4A8 | |
Recombinant protein corresponding to Human MS4A8B. Recombinant protein control fragment (Product #RP-88704). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction