Learn More
MSA™ Advantage™ 1000 CBA-RCA Gas Masks
Clean, simplified respirator is up to 40% lighter than conventional full-face respirators
Manufacturer:  MSA™ 813860
 View more versions of this product
Catalog No. 19-042-091
  Check Availability
Add to cart
Disclaimers:
Note: Canister is effective against OC agents and hydrogen cyanide [HCN], but has not been tested or approved by NIOSH in either of these applications. Advantage mask can be used with canister only if supplied port plug is used to seal opposite port.
Specifications Description & Specifications
Specifications
Gas Mask
Canister, nosecup with two check valve and identification tag
NIOSH, CBRN
Cbrn incidents, civil disturbances, clandestine labs
Small
Dual-canister mount