Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Advantage™ 1000 Full Facepiece Respirators

Full-facepiece respirators are constructed of soft rubber and feature a wraparound flexible urethane lens that offers a comfortable fit with increased vision.

Supplier:  MSA™ 805420

 View more versions of this product

Catalog No. 18-999-1506


Explore available promotions
Add to Cart

Requires : Advantage cartridges and filters (sold separately). Also compatible with Comfo Series cartridges when used with adapter 18-999-1504.

Description

Description

Low-maintenance, weighs 40% less than conventional full-face respirators. Constructed of soft Hycar™™ rubber that provides the feel and comfort of silicone but with better permeation resistance.

  • Fully elastic, six-point head harness adjusts easily
  • Interchangeable Advantage Series cartridges enhance versatility and are compatible with Advantage 200, Advantage 420, Advantage 4000 and Advantage 3200 respirators
  • Unique inturned lip seal on facepiece provides an exceptional seal against a wide range of facial contours
  • Flexible urethane lens is integrally bonded to the facepiece, eliminating the need for rigid lens retainers
  • Includes a speaking diaphragm for clear short-range communications and a nosecup to reduce lens fogging in low temperatures or high humidity

Order Info

Optional lens cover, lens shields, and ESP™ communication system available separately.

Certifications

NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.

Specifications

Specifications

AS NZS 1716:2003
Black
Full Facepiece Respirator
Hycar™
Advantage™ Series cartridges
Speaking diaphragm and nosecup
805420
1.25 lb.
NIOSH 42 CFR Part 84
Hycar Rubber
Large
Fire Service, general industry, oil and gas, utilities
Elastic
Urethane
6-point
0.567 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Accessories
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.