Learn More
MSA™ Advantage™ 3100 Full-Facepiece Respirators
With scratch-resistant, wraparound lens that eliminates distortion and increases peripheral vision
Supplier:  MSA™ 10028999
Catalog No. 19-121-082A
Only null left
Add to Cart
Description
Description
  • With low breathing resistance
  • Standard nosecup reduces fogging
  • Pliable silicone facepiece
  • Pre-adjustable straps for consistent fit
  • Single-port, standard thread filter connection
  • With rubber harness
  • Size: small
Specifications
Specifications
Full Facepiece Respirator
Small
Single port, standard thread filter connection, rubber harness
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.