MSA™ Advantage™ 4000 Facepieces
Manufacturer: MSA™ 10075914
 View more versions of this product
Catalog No. 18-139-400
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Certifications/Compliance ANSI Z87.1 2003, NIOSH
    For Use With (Equipment) Air purifying respirator, supplied air respirator and powered air purifying respirator
    Lens Material Rugged Polycarbonate with hard coating
    Ports Single
    Includes Nosecup, rugged Polycarbonate lens, one-way check valve, push-to-connect adapter, Polyester net head harness with four points of suspension, clear flexible cover lens, instruction manual, black Silicone facepiece blank or elastomers
    Size Small
  • Description & Specifications
Related Products
Product Content Feedback