MSA™ Advantage™ 4000 Facepieces
Manufacturer: MSA™ 10075921
 View more versions of this product
Catalog No. 18-139-409
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance ANSI Z87.1 2003, NIOSH
    For Use With (Equipment) Air purifying respirator, supplied air respirator and powered air purifying respirator
    Lens Material Rugged Polycarbonate with hard coating
    Ports Single
    Includes Nosecup, rugged Polycarbonate lens, one-way check valve, push-to-connect adapter, Polyester net head harness with four points of suspension, clear flexible cover lens, instruction manual, Hycar™ Rubber facepiece blank or elastomers
    Size Large
  • Description & Specifications
Related Products
Product Content Feedback