Learn More
MSA™ Advantage™ 4000 Facepieces
Manufacturer:  MSA™ 10075904
 View more versions of this product
Catalog No. 18-139-406
Add to cart
Specifications
Specifications
ANSI Z87.1 2003, NIOSH
Air purifying respirator, supplied air respirator and powered air purifying respirator
Single
Medium
Rugged Polycarbonate with hard coating
Nosecup, rugged Polycarbonate lens, one-way check valve, push-to-connect adapter, Rubber head harness with four points of suspension, clear flexible cover lens, instruction manual, black Silicone facepiece blank or elastomers