Learn More
MSA™ Advantage™ 4000 Facepieces
Manufacturer:  MSA™ 10083801
 View more versions of this product
Catalog No. 18-139-421
Add to cart
Specifications
Specifications
ANSI Z87.1 2003, NIOSH
Air purifying respirator, supplied air respirator and powered air purifying respirator
Single
Large
Rugged Polycarbonate with hard coating
Nosecup, rugged Polycarbonate lens, one-way check valve, Polyester net harness with four points of suspension, clear flexible cover lens, instruction manual, Hycar™ Rubber facepiece blank or elastomers