Learn More
MSA™ Advantage™ 4000 Facepieces
Manufacturer:  MSA™ 10083803
 View more versions of this product
Catalog No. 18-139-423
Add to cart
Specifications
Specifications
ANSI Z87.1 2003, NIOSH
Air purifying respirator, supplied air respirator and powered air purifying respirator
Single
Large
Rugged Polycarbonate with hard coating
Nosecup, rugged Polycarbonate lens, one-way check valve, Rubber head harness with four points of suspension, clear flexible cover lens, instruction manual, Hycar™ Rubber facepiece blank or elastomers