Learn More
MSA™ Advantage™ 4000 Full Facepiece Respirators, Push-To-Connect Adapter
Full-face reusable respirators with push-to-connect adapter are ideal for a variety of applications. Advantage™ 4000 series is compatible with several MSA respirator models, including air-purifying respirators, supplied-air respirators and powered air-purifying respirators.
Supplier:  MSA™ 10075905
 View more versions of this product
Catalog No. 18-139-407
Add to Cart
Description
Description
  • Single-port facepiece with push-to-connect adapter
  • Compatible with MSA air-purifying respirators, supplied-air respirators and powered air-purifying respirators
  • NIOSH-approval 42 CFR Part 84
  • 4-pt suspension with rubber or polyester net head harness
  • Standand fog-reducing nosecup
  • Facepiece material is available in silicone or Hycar™
  • Three standard sizes to ensure a secure fit
  • Components lock into place in seconds for quick, convenient performance
  • Ideal for use in utilities, mining, oil and gas, fire service, construction, confined space, asbestos abatement, welding and sanding/grinding applications
Specifications
Specifications
ANSI Z87.1 2003
Black
Full Facepiece Respirator with Adapter
Hycar™
Comfo series aerosol filters and chemical cartridges
Push-to-connect Adapter
Polycarbonate
Single
NIOSH 42 CFR Part 84
Hycar™
Medium
Utilities, mining, oil and gas, fire service, construction, confined space
Rubber
Hard coating
10075905
4-point
Product Suggestions
Product Suggestions