Learn More
MSA™ Advantage™ 4000 Twin-Port Facepiece
Two bayonet cartridge adapters mount directly onto the facepiece lens for compatibility with all Advantage cartridges and filters
Manufacturer:  MSA™ 10083783
 View more versions of this product
Catalog No. 18-139-431
Add to cart
Description
Description
  • Approved with all MSA Advantage Cartridges
  • Features two mounting locations for cartridges or filters on the right and left sides
  • Standard nose cup with twin inhalation check valves
Compliance
NIOSH approved under 42 CFR 84 for use against a variety of respiratory hazards when used as designed with appropriate cartridges and filters in conjunction with OSHA fit testing requirement.
Specifications
Specifications
ANSI Z87.1 2003, NIOSH
Respiratory applications
Hycar™ Rubber
Nosecup, bayonet adapters in lens, facepiece elastomers
Rubber
NIOSH 42 CFR Part 84
Msa™ respirator models
Twin-Port Facepiece
Medium
4 point