MSA™ Advantage™ Cartridges

Lightweight for comfort, but designed for maximum protection

Manufacturer: MSA™ 815368

 View more versions of this product

Catalog No. 18-999-3687

  Check Availability
Add to Cart

    Specifications

    Quantity 2 Pack
    Color Olive, Magenta
    For Use With (Equipment) Advantage™ 200 ls and 420 half masks and full facepiece respirator (1000, 3000 and 4000 series)
    Product Type Advantage™ Respirator Cartridge
    For Use With (Application) Asbestos abatement, welding, sanding, grinding, painting, public events, search, rescue
    Mounting Type Bayonet
    Certifications OSHA, NIOSH 42CFR, Part 84, EN 14387, AS NZS 1716, GOST, DSTU
    Cartridge Type Mercury Vapor Cartridge
    Filter Efficiency 99.97%
    Filter Type P100 Filter

    • Particulate, chemical, and combination cartridges
    • Bayonet-style mounting allows easy installation—cartridges lock into place with a single twist
    • Interchangeable between Advantage 200LS, Advantage 420, Advantage 1000, Advantage 4000 and Advantage 3200 respirators

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit