MSA™ AirHawk™ II Self-Contained Breathing Apparatus: Advantage 4000 Model
Lightweight, durable SCBA builds on the success of the established AirHawk air mask by providing options not typically offered in an industrial air mask
Manufacturer: MSA™ 10095800
 View more versions of this product
Catalog No. 17-200-000
  Check Availability
Add to Cart
Alert
Do not use for fire and potential firefighting-related applications. Unit is not NFPA approved for fire service.
    Specifications
    Certifications NIOSH, Non-NFPA
    Harness Style Five strap, Rubber
    For Use With (Application) Suited for use in many industrial IDLH (immediately dangerous to life or health) applications
    Facepiece Type Ultra Elite
    Includes Facepiece: Ultra Elite; Rubber head harness; Hard case
    Material Medium Hycar™
    Product Type SCBA Assembly
    Duration 30 min.
  • Description & Specifications
Related Products
Product Content Feedback