MSA™ AirHawk™ II Self-Contained Breathing Apparatus: Advantage 4000 Model
Lightweight, durable SCBA builds on the success of the established AirHawk air mask by providing options not typically offered in an industrial air mask
Manufacturer: MSA™ 10116781
 View more versions of this product
Catalog No. 17-200-007
  Check Availability
Add to Cart
Alert
Do not use for fire and potential firefighting-related applications. Unit is not NFPA approved for fire service.
    Specifications
    Certifications NIOSH, Non-NFPA
    Facepiece Type Advantage 4000
    Harness Style Nylon with chest strap, rubber
    For Use With (Application) Suited for use in many industrial IDLH (immediately dangerous to life or health) applications
    Includes Advantage 4000 facepiece, Rubber net head harness, Harness carrier: Nylon w/chest strap; Cylinder: carbon - 30 minutes
    Material Medium Hycar™
    Product Type SCBA Assembly
    Duration 30 min.
  • Description & Specifications
Related Products
Product Content Feedback