Learn More
MSA™ ALTAIR™ Pro Single-Gas Detector
Sensor and battery replacement options give this instrument an indefinite service life
Manufacturer:  MSA™ 10076733
 View more versions of this product
Catalog No. 19-100-806
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
3.4 x 2 x 1 in. (8.63 x 5.08 x 2.54cm)
-20° to +50°C
Audible, visual, vibrating
2.54cm
54
8.63cm
0.1%
4 oz.
2 in.
-4° to +122°F
2 Years
1 in.
10 to 95%
3.4 in.
0 to 25%
O2-R
113g
5.08cm