Learn More
MSA™ Comfo™ and Comfo Classic™ Half-Mask Respirators
Available in Hycar™ rubber or silicone, Comfo™ series half-mask respirators feature a unique face seal design for a comfortable fit.
Supplier: MSA™ 808073
Requires : Comfo Series cartridges and filters (see separate listings).
Description
- Protection from exposure to vapors, gases, and particulates
- SoftFeel™ facepiece material increases softness
- Choose from Hycar™ rubber or silicone
- Face seal design provides tight, comfortable fit
- Ideal markets include utilities, oil & gas, construction and general industry
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Specifications
NIOSH 42 CFR Part 84 | |
Silicone | |
Large | |
Utilities, oil and gas, construction, general industry, welding | |
808073 |
Black | |
Half-Mask Respirator | |
Silicone | |
Comfo™ Series cartridges and filters |