Learn More
MSA Detector Tubes
Monitor air contaminants without extractions, analyses, or complicated calculations.
Catalog Number | Mfr. No. | Measurement Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Measurement Range | Price | Quantity & Availability | |||||
18-999-3518
|
MSA
804135 |
10 to 300 |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
18-999-3512
|
MSA
804142 |
1 to 50 |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
18-999-3573
|
MSA
804589 |
1 to 80 |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
MSA/AUER detector tubes are a reliable method of testing for more than 120 hazardous gases and vapors. Detector tubes are flame sealed glass tubes containing treated adsorbent granules that react with a specific compound or group of compounds, causing the adsorbent to change color. A sample is collected by attaching the detector tube to a special bellows-type pump that draws a known volume of air with each stroke. After sampling, the length of adsorbent bed that has undergone the color change is measured from a scale printed on the tube, allowing direct reading of the vapor concentration no extraction, no analysis, and no calculations are required.
Because they are simple to use, detector tubes have been used in a broad range of applications. They are particularly useful when quick screening is required, such as in Haz-Mat Response, and in industrial situations in which specific groups of compounds are monitored regularly.
Order Info
Designed for use with Kwik-Draw Pumps. Also can be used with Toximeter II Automatic Detector Tube pump, Dräger Model 31 Bellows Pump, and Dräger accuro Pump (see individual entries). Tubes come 10 per pack.
Specifications
10 | |
Glass | |
Break off |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.