Learn More
MSA™ Evotech™ Construction Full Body Harnesses
Exceptional comfort, ease of use, durability and user safety
Supplier:  MSA™ 10112708
 View more versions of this product
Catalog No. 17-200-120
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • No binding edge on shoulder padding to prevent neck chafing
  • Breathable padding with moisture-wicking material keeps users cooler
  • Leg strap design keeps leg straps in place to increase mobility and comfort
  • Variable width sub-pelvic webbing provides 50% more surface area for better weight distribution
  • Dual-color webbing (red pattern outside, black inside) simplifies harness donning
  • Web management system conveniently stores excess webbing
  • Positionable lanyard clips
  • RFID enabled to allow easy tracking and inspection
  • Features integral back pad and tool belt with dual-durometer padding for comfort in work positioning applications
Certifications
CSA Z259.10-06
Compliance
Meets OSHA and ANSI Z359.1
Specifications
Specifications
1.75 in. Polyester with NanoSphere™ coating
Full-body subpelvic harness
Back, Side
Qwik-Connect
Subpelvic Full-Body Harness
Tongue-buckle leg strap
Construction
Shoulder padding; Integrap backpad
OSHA, ANSI Z359.1, ANSI A10.32-2004, CSA Z259.10-06
Standard
400 lb.
Product Suggestions
Product Suggestions