MSA™ Evotech™ Standard Full Body Harnesses
Exceptional comfort, ease of use, durability and user safety
Manufacturer: MSA™ 10105942
 View more versions of this product
Catalog No. 17-200-119
  Check Availability
Add to Cart
    Specifications
    Product Type Subpelvic Full-body Harness
    Style Subpelvic full body
    Buckle Style Chest and Leg Strap: Quick-Connect
    D-Ring Location Back; Side
    Webbing Material Polyester
    Capacity (English) 400 lb.
    Includes Manual
    Certifications/Compliance ANSI Z359.1, A10.32, CSA 259.10-06, OSHA
    For Use With (Application) Construction
    Size Standard
  • Description & Specifications
Related Products
Product Content Feedback