MSA™ Faceshield Visors
Fit easily on MSA faceshield frames
Manufacturer: MSA™ 10109107
 View more versions of this product
Catalog No. 19-717-750
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Alert
  • Visors should always be worn with primary eye protection such as Z87.1-2003 high-impact resistant spectacles or goggles
  • Do not use these visors for welding, elevated temperature applications, or for protection against other infrared radiation sources unless it is approved for such applications
  • Do not use for sports, simulated war games, paintball games, or similar activities
  • Inspect visor frequently and replace immediately if worn, scratched, or damaged
  • Do not use near open flame
    Specifications
    Lens Material Propionate
    Lens Tint Clear
    Certifications/Compliance Meets ANSI Z87.1-2003 standards.
    Shape Molded
    Width (English) 15.5 in.
    Length (English) 8 in.
  • Description & Specifications
Related Products
Product Content Feedback