Learn More
MSA™ Faceshield Visors
Fit easily on MSA faceshield frames
Manufacturer:  MSA™ 10017934
 View more versions of this product
Catalog No. 18-320-730
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
  • Visors should always be worn with primary eye protection such as Z87.1-2003 high-impact resistant spectacles or goggles
  • Do not use these visors for welding, elevated temperature applications, or for protection against other infrared radiation sources unless it is approved for such applications
  • Do not use for sports, simulated war games, paintball games, or similar activities
  • Inspect visor frequently and replace immediately if worn, scratched, or damaged
  • Do not use near open flame
Specifications
Specifications
Clear
7.75 in.
Meets ANSI Z87.1-2003 standards.
15.5 in.
Polycarbonate
Flat