Learn More
MSA™ Filters for Advantage™ Cartridges
For use with Advantage Cartridges
Manufacturer:  MSA™ 816661
 View more versions of this product
Catalog No. 19-033-004C
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
50/Pk.
Prefilter
NIOSH
Respirator Cartridge
Advantage™ series respirator
Effective against particulate aerosols free of oil
Threaded
N95 Filter